Branson CPN-916-042 Stainless Steel Support Rack for 2.5 Gal Ultrasonic Cleaner
Representative image only.
Branson CPN-916-042 Stainless Steel Support Rack for 2.5 Gal Ultrasonic Cleaner
Branson | Mfr: CPN-916-042| Item: 0889823
Safely suspends parts off bottom of tank for optimal cleaning
₹39,500.00INR / Each
₹39,500.00INR / Each
Product Description
Key Features
  • Ideal for cleaning small parts Permits sound waves to penetrate directly to the parts
  • For use with 2.5 gal Branson and Cole-Parmer ultrasonic cleaners
More About this Item
These stainless-steel support racks allow total volume of the ultrasonic cleaner tank to be used while keeping parts, fixtures, or containers about ½" above the tank bottom. By preventing parts from touching the bottom of the tank, the racks eliminate possible damage to the unit and help inhibit ultrasonic activity.
They can also be used as countertop drain racks, holding parts above the countertop surface.
Specifications
  • Length (InchShort)11
  • Width (InchShort)8 1/2
  • Height (InchShort)51/64
  • Length (cm)27.94
  • Width (cm)21.59
  • Height (cm)2.032
  • MaterialStainless Steel
  • DescriptionStainless Steel Support Rack for 2.5 Gal Ultrasonic Cleaner