Branson Concentrated Cleaning Solutions
Maximizes cleaning effectiveness of your ultrasonic cleaner
  • Ideal for use with all types of ultrasonic cleaners
  • Safely remove soils, rust, and oxides from a variety of materials
Select the right cleaning solution to maximize the cleaning effectiveness of your ultrasonic cleaner. Cleaning solutions are formulated to deliver superior cleaning performance for a wide variety of cleaning applications.
Please Note: All the cleaning solutions are all biodegradable except for the liquid rust stripper.
69,700.00 - ₹138,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Applications Dilution Volume (Liters)
Availability Pricing
Branson GP-QT General Purpose Caustic Free Alkaline Cleaner, One Quart Bottle 0880111 Mfr # 000-955-014
COMPARE
General purpose 10 to 12%, 11 to 12 pH 1
Call For Price
Branson BC-QT Buffing Compound Remover/Caustic Free Cleaner (tripoli, rouge, lime, diamond tripoli, etc), One Quart Bottle; 12/CS 0889811 Mfr # BC-QT
COMPARE
Tripoli, rouge, lime, diamond tripoli, etc 6-7% by volume, water 0.9
₹70,400.00INR / Case of 12
Branson OR-QT Oxide Remover Cleaner (general soils, fingerprints, cerium oxide, pitch, waxes), One Quart Bottle; 12/CS 0889812 Mfr # OR-QT
COMPARE
General soils, fingerprints, cerium oxide, pitch, waxes 6-7% by volume, water 0.9
₹73,700.00INR / Case of 12
Branson EC-QT Electronic Caustic Free Alkaline Cleaner (oils, resins, & rosins), One Quart Bottle; 12/CS 0889813 Mfr # EC-QT
COMPARE
Oils, resins, & rosins 2-5% by volume, water 0.9
₹138,000.00INR / Case of 12
Branson IS-QT Industrial Strength Caustic Free Alkaline Cleaner (heavy greases/oils/lubricants), One Quart Bottle; 12/CS 0889814 Mfr # IS-QT
COMPARE
Heavy greases, oils, lubricants 8-10% by volume, water 0.9
₹69,900.00INR / Case of 12
Branson MC-1-QT Metal Caustic Free Alkaline Cleaner (displacing/splitting oils/greases from Al, Cu, brass, & steel), One Quart Bottle; 12/CS 0889815 Mfr # MC-1-QT
COMPARE
Displacing, splitting oils, greases from Al, Cu, brass, & steel 7-10% by volume, water 0.9
₹96,800.00INR / Case of 12
Branson MC-2-QT Metal Caustic Free Alkaline Cleaner (emulsifying oils/greases/soils from Fe, SS, Ti, Cu), One Quart Bottle; 12/CS 0889816 Mfr # MC-2-QT
COMPARE
Emulsifying oils, greases, soils from Fe, SS, Ti, Cu 7-10% by volume, water 0.9
₹69,700.00INR / Case of 12
Branson MC-3-QT Metal Caustic Free Alkaline Cleaner (emulsifying & oils/greases/soils from Al and Al alloys), One Quart Bottle; 12/CS 0889817 Mfr # MC-3-QT
COMPARE
Emulsifying oils, greases, soils from Al and Al alloys 7-10% by volume, water 0.9
₹79,400.00INR / Case of 12
Branson LRS-QT Liquid Rust Stripper (mold/equip cleaner removing rust, scaling, paint, & grease), One Quart Bottle; 12/CS 0889818 Mfr # LRS-QT
COMPARE
Mold, rust, scaling, paint, & grease 6-8 oz per gallon, water 0.9
₹71,900.00INR / Case of 12
Showing 1- 9 of 9