Branson GP-QT General Purpose Caustic Free Alkaline Cleaner, One Quart Bottle
Representative image only.
Branson GP-QT General Purpose Caustic Free Alkaline Cleaner, One Quart Bottle
Branson | Mfr: 000-955-014| Item: 0880111
Maximizes cleaning effectiveness of your ultrasonic cleaner
Call For Price
Call For Price
Buy It Together
Current Item
Branson GP-QT General Purpose Caustic Free Alkaline Cleaner, One Quart Bottle
Branson GP-QT General Purpose Caustic Free Alkaline Cleaner, One Quart Bottle
0880111
Call For Price
+ + + +
Elmasonic Ultrasonic Cleaner Basket, E+ Series; 0.25 gal.
5998835
₹16,400.00INR / Each
+ + =
Total Price 809400.00 ADD 7 ITEMS TO CART
Product Description
Key Features
  • Ideal for use with all types of ultrasonic cleaners
  • Safely remove soils, rust, and oxides from a variety of materials
  • Specially formulated for maintenance cleaning where soil levels are relatively light
  • Biodegradable and caustic free alkaline cleaner
  • 1 year warranty
More About this Item
Select the right cleaning solution to maximize the cleaning effectiveness of your ultrasonic cleaner. Cleaning solutions are formulated to deliver superior cleaning performance for a wide variety of cleaning applications.
Please Note: All the cleaning solutions are all biodegradable except for the liquid rust stripper.
1 Year Warranty
Specifications
  • FormatLiquid
  • Volume (gallons)0.25
  • Volume (Liters)1
  • Dilution10 to 12%, 11 to 12 pH
  • ApplicationsGeneral purpose
  • DescriptionGeneral-purpose ultrasonic cleaning solution, 1 qt bottle