Branson CH Series Ultrasonic Heated Transducerized Cleaning Tanks with GCX Generator
Precise control of ultrasonic energy provides greater cleaning power
  • Maintains solution temperature up to 76 °C (170 °F)
  • Bottom-mounted positive drain connection
  • Fully shielded internal components
  • Generator offers user-friendly digital controls
These ultrasonic heated cleaning tanks are constructed throughout with 316L bright-annealed stainless steel allowing you to use alkaline, mildly acidic, or neutral aqueous solutions. The tanks are able to maintain solution temperatures up to 76 °C (170 °F). All the tanks have drains and fills.
Included with the tanks is the GCX ultrasonic generator which delivers ultrasonic energy in a wide range of frequencies. For precise control of your cleaning process and performance, the compact generator features auto tuning, line/load regulation, and sweep frequency. The user-friendly touchscreen human/machine interface (HMI) offers a wide range of precise, digital settings as well as easy access to system data and setup. A fully variable power control allows you to vary the cavitational intensity from 20% to 100%.
The generator also includes multi-language settings (English, Chinese, and Japanese), power modulation mode which produces four times the peak power in the bath, dual I/O interface with a built-in simple on/off control, and safety features to protect from over-temperature, overload, open circuit, and short circuit
2,844,000.00 - ₹5,484,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Height (InchShort) Width (InchShort)
Availability Pricing
Branson CH1012-40-12 Ultrasonic Heated Transducerized Cleaning Tank, 5 Gal, 10"x12", 40kHz , with GCX Generator 0889837 Mfr # CH1012 BUNDLE
COMPARE
5 16 14
₹2,844,000.00INR / Each
Branson CH1216-40-18 Ultrasonic Heated Transducerized Cleaning Tank, 10 Gal, 12"x16", 40kHz, with GCX Generator 0889838 Mfr # CH1216 BUNDLE
COMPARE
10 20 16
₹3,597,000.00INR / Each
Branson CH1620-40-24 Ultrasonic Heated Transducerized Cleaning Tank, 20 Gal, 16"x20", 40kHz, with GCX Generator 0889839 Mfr # CH1620 BUNDLE
COMPARE
20 24 20
₹4,139,000.00INR / Each
Branson CH2024-40-36 Ultrasonic Heated Transducerized Cleaning Tank, 40 Gal, 20"x24", 40kHz, with GCX Generator 0889840 Mfr # CH2024 BUNDLE
COMPARE
40 28 24
₹5,484,000.00INR / Each