InnoCal NIST-Traceable Calibration; Ultrasonic Cleaner
Representative image only.
InnoCal NIST-Traceable Calibration; Ultrasonic Cleaner
InnoCal | Item: 1705005
₹37,400.00INR / Each
₹37,400.00INR / Each
Buy It Together
Current Item
InnoCal NIST-Traceable Calibration; Ultrasonic Cleaner
InnoCal NIST-Traceable Calibration; Ultrasonic Cleaner
1705005
₹37,400.00INR / Each
+
Cavins CM-3-100 Ultrasonic Cavitation Meter
0889460
₹537,000.00INR / Each
=
Total Price 574400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification
  • EPA, USDA, ISO and GLP/GMPs standards require regular NIST-Traceable calibration of all test equipment being used in critical applications
More About this Item
All measurements reported are traceable to SI units via national standards maintained by NIST and performed in compliance with ANSI Z540, ISO17025 and MIL-STD-45662A. See the sample certificate under the “More Info” section to see how InnoCal’s certificates provide more information in a concise manner than the rest of the industry.
InnoCal is accredited to ISO/IEC 17025 by the American Association for Laboratory Accreditation (A2LA) under certificate number 1746.01. This accreditation is your assurance that the calibration is performed in a qualified, controlled laboratory which is technically proficient and committed to quality and customer-focused continuous improvement. Our scope of A2LA accreditation can be found under the “More Info” section.
Certification on ultrasonic cleaners is performed using NIST-traceable instruments to measure the total power in the cleaner as well as performing industry standard efficacy tests to ensure that the ultrasonic cleaner is operating according to specifications. If the ultrasonic cleaner has a timer or temperature display, the accuracy of those will be compared to NIST-traceable standards.
Specifications
  • Calibration TypeLiquid Testing
  • Instrument Only CalibrationYes
  • System Calibration (Instrument and Probe)Yes
  • DescriptionNIST-Traceable Calibration; Ultrasonic Cleaners