Bradley Portable Pressurized Eyewash with Drench Hose
Goes anywhere to provide onsite emergency relief for eyes and face.
Comes with 8-ft (2.4 m) yellow, reinforced thermoplastic drench hose designed to supply supplemental water to face and body. Stainless steel tank features tire filler valve and 0 to 160 psi air guage. Pressure relief valve prevents accidental over-pressurization.
Flow rates: eyewash exceeds minimum 0.4 gpm at 30 psi; drench hose exceeds minimum 3 gpm at 30 psi.
Compliance: ANSI Z358.1-2004
316,000.00 - ₹449,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Bradley S19-672 Portable Pressurized Eyewash with Drench Hose, 5 gallon 8600008 Mfr # S19-672
COMPARE
Portable Pressurized Eyewash with Drench Hose, 5 gallon
₹316,000.00INR / Each
Bradley S19-690 Portable Pressurized Eyewash with Drench Hose, 10 gallon 8600010 Mfr # S19-690
COMPARE
Portable Pressurized Eyewash with Drench Hose, 10 gallon
₹426,000.00INR / Each
Bradley S19-788 Portable Pressurized Eyewash with Drench Hose, 15 gallon 8600012 Mfr # S19-788
COMPARE
Portable Pressurized Eyewash with Drench Hose, 15 gallon
₹449,000.00INR / Each