Bradley S19-690 Portable Pressurized Eyewash with Drench Hose, 10 gallon
Representative image only.
Bradley S19-690 Portable Pressurized Eyewash with Drench Hose, 10 gallon
Bradley | Mfr: S19-690| Item: 8600010
Goes anywhere to provide onsite emergency relief for eyes and face.
₹426,000.00INR / Each
₹426,000.00INR / Each
Buy It Together
Current Item
Bradley S19-690 Portable Pressurized Eyewash with Drench Hose, 10 gallon
Bradley S19-690 Portable Pressurized Eyewash with Drench Hose, 10 gallon
8600010
₹426,000.00INR / Each
+ =
Total Price 502700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Comes with 8-ft (2.4 m) yellow, reinforced thermoplastic drench hose designed to supply supplemental water to face and body. Stainless steel tank features tire filler valve and 0 to 160 psi air guage. Pressure relief valve prevents accidental over-pressurization.
Flow rates: eyewash exceeds minimum 0.4 gpm at 30 psi; drench hose exceeds minimum 3 gpm at 30 psi.
Compliance: ANSI Z358.1-2004
1 Year Warranty
Specifications
  • Eyewash Fixture MaterialChrome-plated brass
  • Hose MaterialThermoplastic
  • Tank MaterialStainless steel
  • MountPortable
  • ComplianceANSI Z358.1-2009 as a supplemental eyewash only
  • DescriptionPortable Pressurized Eyewash with Drench Hose, 10 gallon