Bradley HALO Eye Wash Bowls
No struggling to activate the eyewash – broad hand flag makes activation easy!
  • Eye wash bowls provide excellent face wash down coverage
HALO eyewashes provide rapid relief to individuals who have been injured by chemicals or particulates. Ergonomic hand activation is easy to reach from both sides of the bowl. Integral strainer reduces debris in the supply water for further eye protection. Bowls are contoured for superior draining, reducing the risk of bacterial growth when not in use. Separate supply and waste pipes meet CEN and plumbing requirements. Supply connection is 1/2" NPT(F); waste connection is 1-1/4" NPT(M).
Compliance: ANSI Z358.1-2009
75,800.00 - ₹127,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bowl Material Mount Description
Availability Pricing
Bradley S19224B Eyewash, wall mount, Stainless steel 8600121 Mfr # S19224B
COMPARE
Stainless steel Wall Mount Eyewash, wall mount, Stainless steel
₹99,300.00INR / Each
Bradley S19214EW Eyewash, Pedestal mount, ABS plastic 8600122 Mfr # S19214EW
COMPARE
ABS plastic Pedestal Mount Eyewash, Pedestal mount, ABS plastic
₹75,800.00INR / Each
Bradley S19214B Eyewash, Pedestal mount, Stainless steel 8600123 Mfr # S19214B
COMPARE
Stainless steel Pedestal Mount Eyewash, Pedestal mount, Stainless steel
₹127,000.00INR / Each