Bradley S19214EW Eyewash, Pedestal mount, ABS plastic
Representative image only.
Bradley S19214EW Eyewash, Pedestal mount, ABS plastic
Bradley | Mfr: S19214EW| Item: 8600122
No struggling to activate the eyewash – broad hand flag makes activation easy!
₹75,800.00INR / Each
₹75,800.00INR / Each
Product Description
Includes
universal eyewash sign and inspection tag.
Key Features
  • Eye wash bowls provide excellent face wash down coverage
  • 1 year warranty
More About this Item
HALO eyewashes provide rapid relief to individuals who have been injured by chemicals or particulates. Ergonomic hand activation is easy to reach from both sides of the bowl. Integral strainer reduces debris in the supply water for further eye protection. Bowls are contoured for superior draining, reducing the risk of bacterial growth when not in use. Separate supply and waste pipes meet CEN and plumbing requirements. Supply connection is 1/2" NPT(F); waste connection is 1-1/4" NPT(M).
Compliance: ANSI Z358.1-2009
1 Year Warranty
Specifications
  • Eyewash Fixture MaterialABS plastic
  • Bowl MaterialABS plastic
  • MountPedestal Mount
  • Outlet Connections1-1/4" NPT(M)
  • Inlet Connection1/2" NPT(F)
  • ComplianceANSI Z358.1-2009
  • DescriptionEyewash, Pedestal mount, ABS plastic