Bio-Simplex Carboys with Handles, PP
  • Slender ergonomic design takes up less space
These carboys are ideal for advanced liquid handling and as a solution storage for several applications.
The design features large top handles for full hand grip and bottom handles for secure grip when holding and transporting. Closed cap technology provides leak-proof top connections. Wide mouth allows easy filling and cleaning. They also feature easy-to-read certified graduation marks with ± 5% accuracy.
Carboys are manufactured from high-quality polypropylene (PP), which is resistant to chemicals and heating up to 135°C. Autoclavable.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Capacity (L) Description
Availability Pricing
Bio-Simplex DCB0580 5 Liter Carboy with Cap WW-15944-10 Mfr # DCB0580
COMPARE
1.3 5
Call For Price
Bio-Simplex DCB1080 10 Liter Carboy with Cap WW-15944-11 Mfr # DCB1080
COMPARE
2.6 10
Call For Price
Bio-Simplex DCB20120 20 Liter Carboy with Cap WW-15944-12 Mfr # DCB20120
COMPARE
5.2 20
Call For Price
Bio-Simplex DCB40120 40 Liter Carboy with Cap WW-15944-13 Mfr # DCB40120
COMPARE
10.5 40