Bio-Simplex DCB1080 10 Liter Carboy with Cap
Representative image only.
Bio-Simplex DCB1080 10 Liter Carboy with Cap
Bio-Simplex | Mfr: DCB1080| Item: 1594411
Call For Price
Call For Price
Product Description
Key Features
  • Slender ergonomic design takes up less space
More About this Item
These carboys are ideal for advanced liquid handling and as a solution storage for several applications.
The design features large top handles for full hand grip and bottom handles for secure grip when holding and transporting. Closed cap technology provides leak-proof top connections. Wide mouth allows easy filling and cleaning. They also feature easy-to-read certified graduation marks with ± 5% accuracy.
Carboys are manufactured from high-quality polypropylene (PP), which is resistant to chemicals and heating up to 135°C. Autoclavable.
Specifications
  • AutoclavableYes
  • MaterialPolypropylene(PP)
  • ColorGray
  • Capacity (Gal)2.6
  • Capacity (L)10
  • Max Temperature (° F)275