BEVCO Laboratory Sit & Stand Stools
Designed for mobility, safety and comfort
“Sit-Stand” backless stool gives support while sitting or standing. Allows worker to stay in upright position while placing weight directly on seat. Ergonomically designed for mobility without putting added pressure on back and lower body of worker. Polyurethane seat has tilt adjustment. Sit-stand stools have pneumatic seat height adjustmen and pneumatic cylinder has a lifetime guarantee. Stools have 360°swivel. The texture on the seat provides a non-slip surface that is puncture-resistant and easy-to-clean.
77,600.00 - ₹83,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Material Seat height (in)
Availability Pricing
Bevco 3505 Sit-stand stool, 22"-32" height, black reinforced plastic base 4762164 Mfr # 3505
COMPARE
22 Plastic 22 to 32
₹77,600.00INR / Each
Bevco 3555 Sit-stand stool, 22"-32" height, polished aluminum base 4762166 Mfr # 3555
COMPARE
22 Aluminum 22 to 32
₹83,600.00INR / Each