Benchmark Scientific BeadBug™ 3 Microtube Homogenizer
Powerful disruptive mixing for multiple samples
  • Sealed homogenization prevents cross-contamination
  • Quick and effective personal-size homogenizer grinds and lyses soft cells to tough materials
  • Process up to 3 samples simultaneously to increase throughput
  • Timer: 3 seconds to 3 minutes
These microtube bead homogenizers allow you to process one, two, or three 2.0-mL samples at the same time, usually within 45 seconds. Constant high-velocity impacts from hardened microbeads probides rapid cell and tissue disruption, even for tough, fibrous, or hard tissues. The small footprint allows you to keep the homogenizer close by for more efficient sample preparation. Timer function gives you the freedom to step away during processing to work on other tasks.
Prefilled disposable tubes with screw caps make the process even easier. Empty tubes and bulk beads are also available for customization and optimization. Choose from a variety of bead materials and sizes for the best fit to your application.
229,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Sample Capacity (mL) Speed (rpm)
Availability Pricing
Benchmark Scientific BeadBug 3 Microtube Homogenizer, 115 VAC 3627102 Mfr # D1030
COMPARE
115 2 2800
₹229,000.00INR / Each
Benchmark Scientific BeadBug 3 Microtube Homogenizer, 230 VAC 3627103 Mfr # D1030-E
COMPARE
230 2 4000
₹229,000.00INR / Each