Belly Button Shaker
Compact design is ideal for crowded incubators
  • Adjustable-angle platform is flexibly anchored at four corners, making an effective combination of orbital shaker and platform mixer
  • Wide ambient operating range of 32 to 140°F (0 to 60°C) for use in incubators and cold rooms
Small footprint takes up little space in the lab and fits inside incubators, ovens, and refrigerated units without having to rearrange shelves or disturb samples. This shaker is ideal for cell culture and tissue culture applications — the undulating motion and slow speeds provide superior growth conditions without shearing cells. The easily adjustable speed and platform pitch angle controls provide a wide range of agitation conditions, from extremely gentle to vigorous.
Brushless motor provides cool, quiet operation and allows for operation at very slow speeds without cogging. Shaker features lightweight durable construction, variable speed control, and a flat power cord that fits easily through incubators doors without compromising the environment inside.
265,000.00 - ₹282,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Ibi Scientific BBUAAUV1S Belly Button Shaker, 0 to 100 rpm, 94 to 240 VAC WW-51650-06 Mfr # BBUAAUV1S
COMPARE
Belly Button Shaker, 0 to 100 rpm, 94 to 240 VAC
₹265,000.00INR / Each
Ibi Scientific BBULS0001 Belly Button Shaker with Finger Mat, Universal Voltage WW-51650-07 Mfr # BBULS0001
COMPARE
Belly Button Shaker with Finger Mat, Universal Voltage
₹282,000.00INR / Each