Beaker / Bottle / Flask Brushes
Contoured brushes are designed to clean hard-to-reach areas
Radial tip brushes allow you to remove difficult deposits without scratching your container. Folding tip and double-tufted brushes are made to fold flat through the bottle neck and expand to fill the container. Angle head brushes are manufactured to angle the brush along the curvature of the container with minimal strain. Volumetric brushes feature an angled handle which bends to fit the container shape for cleaning pear-shaped, Erlenmeyer, round bottom, flat bottom, Nephelo, Kjeldahl, filtering, and volumetric flasks.
11,400.00 - ₹28,000.00INR / Pkg of 12
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Handle Length (in) Material
Availability Pricing
COMPARE
White 6 Natural bristles, wire handle
₹11,400.00INR / Pkg of 12
COMPARE
Black 7.5 Natural bristles, wire handle
₹11,800.00INR / Pkg of 12
COMPARE
Black 19.5 Natural bristles, wire handle
₹16,200.00INR / Pkg of 12
COMPARE
White 12 Nylon bristles, plastic handle
₹16,400.00INR / Pkg of 12
COMPARE
White 10 Nylon bristles, plastic handle
₹28,000.00INR / Pkg of 12