Backsaver Soil Sampling Handles
Choose from five different handle styles each designed for sampling to different depths
  • Use the basic handle when sampling soft soils
  • All handles are made of nickel-plated carbon steel for long life
The shallow, medium, and deep backsaver handles are designed with a foot pedal to drive the sampler using your body weight and gravity (see specifications information below for specific depth of each handle). Complete your soil sampler by ordering sampling tubes and extensions (sold separately). Use and optional extension rod for increased depth.
28,000.00 - ₹177,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Material Max Sampling Depth (in) Description
Availability Pricing
COMPARE
Nickel-plated carbon steel 42 Soil Sampling Basic Handle; 36" to 42" Sampling Depth
₹28,000.00INR / Each
COMPARE
Nickel-plated carbon steel 18 Soil Sampling Shallow Backsaver Handle; 12" to 18" Sampling Depth
₹79,500.00INR / Each
COMPARE
Nickel-plated carbon steel 46 Soil Sampling Medium Backsaver Handle; 18" to 46" Sampling Depth
₹151,000.00INR / Each
COMPARE
Nickel-plated carbon steel 66 Soil Sampling SDeep Backsaver Handle; 18" to 66" Sampling Depth
₹177,000.00INR / Each