Bacharach Fyrite InTech Combustion Analyzer
Economical combustion analyzer with great features
The Fyrite® InTech™ combustion analyzer is ideal for use on residential and light commercial furnaces and appliances. Take accurate measurements of oxygen, stack temperature, and ambient temperature. The high-contrast, backlit LCD lets you see your results even in dimly lit areas. Front-panel arrow and function buttons allow easy navigation of the menu system to select parameter and display options. The B-Smart® sensor technology allows precalibrated CO sensor replacement in the field (order replacement sensors separately).
  • Works with natural gas, oil #2, oil #6, kerosene, propane, and biodiesel 5% (B5)
  • Calculate CO2, CO (air free), and excess air
  • Store up to 10 records for recall on the display
  • Easily navigate the menu system through the function and arrow buttons
  • Time-and-date stamp your test results
  • Auto power off and sensor purge feature
  • Select the display language from English, French, or Spanish
224,000.00 - ₹366,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Includes
Availability Pricing
Bacharach Fyrite Intech Combustion Analyzer, O2 measurement 8649570 Mfr # H0024-8510
COMPARE
O2 sensor, soft case
₹224,000.00INR / Each
Bacharach Fyrite Intech Combustion Analyzer, O2/CO measurement 8649571 Mfr # H0024-8511
COMPARE
O2/CO sensor, soft case
₹280,000.00INR / Each
Bacharach Fyrite Intech Combustion Analyzer; O2/CO, carrying case 8649572 Mfr # H0024-8523
COMPARE
O2/CO sensors, rubber boot, hard case
₹311,000.00INR / Each
Bacharach Fyrite Intech Combustion Analyzer; O2/CO, Reporting Kit 8649573 Mfr # H0024-8512
COMPARE
O2/CO sensors, reporting lit, rubber boot, hard case
₹366,000.00INR / Each