Azlon Heavy-Duty Tweezers, PMP
Heavy-duty, readjusting tweezers are available in sharp or rounded ends with fine serrated grips. Tweezers are made from rigid, impact-resistant and temperature-resistant polymethylpentene (PMP). Able to withstand temperatures up to 392°F (200°C) for short periods and 356°F (180°C) continuously. PMP is ideal for use with diluted and concentrated acids, alcohols, aldehydes, bases, esters, mineral oils, and vegetable oils. Autoclavable.
8,200.00 - ₹24,400.00INR / Case of 6
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Length (in)
Availability Pricing
Azlon 516555-0001 Heavy-Duty PMP Serrated Pointed Fine Tip Tweezers, 4.5" L; 6/Cs WW-07290-50 Mfr # 516555-0001
COMPARE
4 1/2
₹8,200.00INR / Case of 6
Azlon 516555-0004 Heavy-Duty PMP Serrated Rounded Tip Tweezers, 9.75" L; 6/Cs WW-07290-56 Mfr # 516555-0004
COMPARE
9 3/4
₹24,400.00INR / Case of 6