Axygen® Polypropylene Centrifuge Tubes
Reliable conical centrifuge tubes for high-performance sample processing and storage
  • 15 mL and 50 mL conical tubes engineered for high-speed workflows up to ~12,000 x g
  • DNase/RNase-free, nonpyrogenic construction supports sensitive molecular and cell biology applications
  • Graduated polypropylene tubes with secure screw caps for reliable handling, storage, and transfer
  • Available in pack and case configurations
The Axygen® centrifuge tube line delivers dependable performance for everyday laboratory workflows, from routine sample prep to high-throughput processing. With both 15 mL and 50 mL conical formats available, these tubes are designed for compatibility across standard centrifugation systems while maintaining sterility and mechanical integrity under high g-forces. Multiple packaging options for a variety of application needs.
4,100.00 - ₹85,500.00INR / Case of 500
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Packaging Sterile Tube Capacity (mL)
Availability Pricing
Axygen 15 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Bulk Packed, 50/Sleeve, 500/Case 4846001 Mfr # SCT-15ML-500
COMPARE
Case of 500 Yes 15
₹33,700.00INR / Case of 500
Axygen 15 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Bulk Packed, Sterile, 25/Sleeve, 500/Case 4846002 Mfr # SCT-15ML-25-S
COMPARE
Case of 500 Yes 15
₹45,900.00INR / Case of 500
Axygen 15 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Racked, Sterile, 50/Rack, 500/Case 4846003 Mfr # SCT-15ML-R-S
COMPARE
Case of 500 Yes 15
₹51,900.00INR / Case of 500
Axygen 50 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Bulk Packed, 50/Sleeve, 500/Case 4846004 Mfr # SCT-50ML-500
COMPARE
Case of 500 Yes 50
₹54,300.00INR / Case of 500
Axygen 50 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Bulk Packed, Sterile, 25/Sleeve, 500/Case 4846005 Mfr # SCT-50ML-25-S
COMPARE
Case of 500 Yes 50
₹64,100.00INR / Case of 500
Axygen 50 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Racked, Sterile, 25/Rack, 500/Case 4846006 Mfr # SCT-50ML-R-S
COMPARE
Case of 500 Yes 50
₹85,500.00INR / Case of 500
Axygen 15 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Bulk Packed, 50/Sleeve 4846032 Mfr # SCT-15ML-500
COMPARE
Sleeve of 50 Yes 15
₹4,100.00INR / Pkg of 50
Axygen 15 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Bulk Packed, Sterile, 25/Sleeve 4846033 Mfr # SCT-15ML-25-S
COMPARE
Sleeve of 25 Yes 15
₹5,400.00INR / Pkg of 25
Axygen 15 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Racked, Sterile, 50/Rack 4846034 Mfr # SCT-15ML-R-S
COMPARE
Rack of 50 Yes 15
₹6,000.00INR / Pkg of 50
Axygen 50 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Bulk Packed, 50/Sleeve 4846035 Mfr # SCT-50ML-500
COMPARE
Sleeve of 50 Yes 50
₹4,600.00INR / Pkg of 50
Axygen 50 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Bulk Packed, Sterile, 25/Sleeve 4846036 Mfr # SCT-50ML-25-S
COMPARE
Sleeve of 25 Yes 50
₹6,300.00INR / Pkg of 25
Axygen 50 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Racked, Sterile, 25/Rack 4846037 Mfr # SCT-50ML-R-S
COMPARE
Rack of 25 Yes 50
₹7,000.00INR / Pkg of 25
Showing 1- 12 of 12