Axygen 15 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Racked, Sterile, 50/Rack, 500/Case
Representative image only.
Axygen 15 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Racked, Sterile, 50/Rack, 500/Case
| Mfr: SCT-15ML-R-S| Item: 4846003
Reliable conical centrifuge tubes for high-performance sample processing and storage
₹51,900.00INR / Case of 500
₹51,900.00INR / Case of 500
Product Description
Key Features
  • 15 mL and 50 mL conical tubes engineered for high-speed workflows up to ~12,000 x g
  • DNase/RNase-free, nonpyrogenic construction supports sensitive molecular and cell biology applications
  • Graduated polypropylene tubes with secure screw caps for reliable handling, storage, and transfer
  • Available in pack and case configurations
More About this Item
The Axygen® centrifuge tube line delivers dependable performance for everyday laboratory workflows, from routine sample prep to high-throughput processing. With both 15 mL and 50 mL conical formats available, these tubes are designed for compatibility across standard centrifugation systems while maintaining sterility and mechanical integrity under high g-forces. Multiple packaging options for a variety of application needs.
Specifications
  • Max RCF x g12000
  • SterileYes
  • Tube Capacity (mL)15
  • Tube MaterialPolypropylene
  • Tube ColorClear
  • Tube Bottom ShapeConical
  • Cap IncludedYes
  • Cap StyleScrew cap
  • Cap MaterialHigh density polyethylene
  • GraduationsYes
  • PackagingCase of 500
  • Description15 mL Polypropylene (PP) Centrifuge Tubes, Conical Bottom with Screw Cap, Racked, Sterile, 50/Rack, 500/Case