Ashcroft T4 Series Temperature Switches
Rugged—reliable in harsh environments
  • Quick, accurate response via SAMA Class II vapor pressure thermal system
  • Choose the switch to meet your needs—wide variety of lengths and temperature ranges available
  • Easy to install—watertight enclosure meets NEMA 3, 4, 4X, 13, and IP66 requirements
Single set point, fixed deadband, SPDT switches feature 316 stainless steel systems for long-lasting performance—even in harsh environments. Choose direct-mounting switches with rigid probes—great for use with 3/8" nominal bore thermowells in areas with high vibration or pressure. Remote-mount switches have spring-mounted capillaries with 302 stainless steel armor and include a 1/2" NPT(M) bushing adapter.
122,000.00 - ₹141,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Ashcroft T4 Temperature Switch, Direct Mount, 2.75"L Bulb; 75 to 205°F 3901212 Mfr # T4 24 TS-027 75/205F
COMPARE
Temperature Switch, Direct Mount, 2.75"L Bulb; 75 to 205°F
₹122,000.00INR / Each
Ashcroft T4 Temperature Switch, Direct Mount, 4.0"L Bulb; 0 to 100°F 3901221 Mfr # T4 24 TS-040 0/100F
COMPARE
Temperature Switch, Direct Mount, 4.0"L Bulb; 0 to 100°F
₹122,000.00INR / Each
Ashcroft T4 Temperature Switch, Remote Mount, 5'; 75 to 205°F 3901244 Mfr # T4 24 T05-030 XBX 75/205F
COMPARE
Temperature Switch, Remote Mount, 5'; 75 to 205°F
₹141,000.00INR / Each