Ashcroft Pressure Snubbers
Protect your gauges from pressure surges and spikes
  • Reduces pressure spikes only
  • Spikes last for mere milliseconds with a snubber
2,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Media Compatibility
Availability Pricing
Ashcroft BW41 Pressure Snubber for Air, Brass, 1/4" NPT(M)/(F), 1500 psi 6843112 Mfr # BW41
COMPARE
Air
₹2,300.00INR / Each
Ashcroft BW42 Pressure Snubber for Water-Light Oil, Brass, 1/4" NPT(M)/(F), 1500 psi 6843113 Mfr # BW42
COMPARE
Water to light oil
₹2,300.00INR / Each