Ashcroft BW42 Pressure Snubber for Water-Light Oil, Brass, 1/4" NPT(M)/(F), 1500 psi
Representative image only.
Ashcroft BW42 Pressure Snubber for Water-Light Oil, Brass, 1/4" NPT(M)/(F), 1500 psi
Ashcroft | Mfr: BW42| Item: 6843113
Protect your gauges from pressure surges and spikes
₹2,300.00INR / Each
₹2,300.00INR / Each
Product Description
Key Features
  • Reduces pressure spikes only
  • Spikes last for mere milliseconds with a snubber
  • Compatible with water-light oil
  • 1 year warranty
More About this Item
Snubbing element consists of a 3/8" dia x 1/8" thick
micro metallic stainless steel filter.
1 Year Warranty
Specifications
  • Max Pressure (PSI)1500
  • Wetted MaterialsBrass with 3/8" dia × 1/8" thick micro stainless steel filter
  • Process Connection1/4" NPT(M)/1/4" NPT(F)
  • Media CompatibilityWater to light oil
  • DescriptionPressure Snubber with 1/8" Filter, Brass, 1/4" NPT(M)/(F), 1500 psi