Ashcroft Economical Differential Pressure Gauges
Excellent for filtration systems, leak monitoring, and maintaining flow and tank levels
  • Close-coupled magnetic rack and pinion design achieves superior stability.
  • Weather-resistant stainless steel case, aluminum body, and Buna N seals
  • Use glycerin filled gauges in high vibration areas to achieve reliable and stable readings.
  • TrueZero™ indication for dependable pressure readings at all times within ±2% full-scale accuracy (ascending)
68,700.00 - ₹92,400.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Pressure (PSID)
Availability Pricing
Ashcroft 11302.5 2-½" Economical Differential Pressure Gauges, 0 to 15 psid 6840300 Mfr # 25-1130-FD-25S-15#
COMPARE
±2% full-scale (ascending) 15
₹68,700.00INR / Each
Ashcroft 11302.5 2-½" Economical Differential Pressure Gauges, 0 to 20 psid 6840302 Mfr # 25-1130-FD-25S-20#
COMPARE
±2% full-scale (ascending) 20
₹68,700.00INR / Each
Ashcroft 11302.5 2-½" Economical Differential Pressure Gauges, 0 to 30 psid 6840304 Mfr # 25-1130-FD-25S-30#
COMPARE
±2% full-scale (ascending) 30
₹68,700.00INR / Each
Ashcroft 11302.5 2-½" Economical Differential Pressure Gauges, 0 to 100 psid 6840308 Mfr # 25-1130-FD-25S-100#
COMPARE
±2% full-scale (ascending) 100
₹68,700.00INR / Each
Ashcroft 11302.5 Glycerine Filled Differential Gauge, 0 to 15psid 6840310 Mfr # 25-1130-FD-L-25S-15#
COMPARE
±2% full-scale (ascending) 15
₹92,400.00INR / Each
Ashcroft 11302.5 Glycerine Filled Differential Gauge, 0 to 30 psid 6840314 Mfr # 25-1130-FD-L-25S-30#
COMPARE
±2% full-scale (ascending) 30
₹92,400.00INR / Each
Ashcroft 11302.5 Glycerine Filled Differential Gauge, 0 to 100 psid 6840318 Mfr # 25-1130-FD-L-25S-100#
COMPARE
±2% full-scale (ascending) 100
₹92,400.00INR / Each
Showing 1- 7 of 7