Ansell Endurosaf Aprons
Lightweight protection from chemicals, oils, and abrasion
These smooth, lightweight aprons are made of ENDURO 2000™ material that gives them excellent chemical, oil, and abrasion resistance. Easy-to-clean aprons maintain their shape and stay wrinkle-free even after extended wear and fluctuating temperatures. Aprons are 8-mil thick and have 1" wide adjustable neckstrap and side-ties.
3,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Width (in)
Availability Pricing
Ansell 56-800 SAF-45W 951922 Apron, White, 35" W x 45" L; 1/Pack WW-81620-01 Mfr # 56-800 SAF-45W 951922
COMPARE
White 45 35
₹3,600.00INR / Each
Ansell 56-802 SAF-45B 951928 Apron, Blue, 35" W x 45" L; 1/Pack WW-81620-05 Mfr # 56-802 SAF-45B 951928
COMPARE
Blue 45 35
₹3,600.00INR / Each