AEMC 1050, 1060, 5050, 5060 1000 and 5000V Digital Megohmmeters
Automated, user-friendly system for testing insulation and resistance
  • Provides direct measurement of dielectric, polarization index, and capacitance.
Measure insulation voltage levels up to 1000 or even to 5000 V. The programmable memory stores resistance values for specified time intervals. The backlit LCD features a dual display with a digital measurement and a bar graph to monitor changes or trends in readings. Automatic test inhibitor shuts down meter when live circuits exceed 25V — protecting the operator and equipment. The rugged case features a weatherproof design for heavy duty conditions.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Interface Max Resistance (G ohms) Max Voltage (Voltage)
Availability Pricing
AEMC 1060 Industrial Digital Megohmmeter w/RS-232, 1000 V 2744306 Mfr # 1060
COMPARE
RS-232 4000 1000
Call For Price
AEMC 5050 Industrial Digital Megohmmeter, 5000 V 2744310 Mfr # 2130.20
COMPARE
None 10000 5000
Call For Price
AEMC 5060 Industrial Digital Megohmmeter w/RS-232, 5000 V 2744312 Mfr # 2130.21
COMPARE
RS-232 10000 5000