Advantec Stainless Steel Filter Holders
Open holders without disconnecting lines
  • Bodies and perforated support screens are 304 SS
  • PTFE gaskets
  • Silicone o-rings
  • Autoclave with membrane in place
Syringe holders have luer fittings. In-line holders have 1/4" NPT(F) and 1/4" NPT(M) ports. Supplied with 3/8" hose barb connectors and two perforated 304 stainless steel support screens.
21,400.00 - ₹155,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Body Material Filter Diameter (mm) Inlet Connection
Availability Pricing
Advantec 301000 Stainless Steel Filter Holder, Syringe; 13mm dia WW-02928-10 Mfr # 301000
COMPARE
Stainless Steel 13 Female luer
₹21,400.00INR / Each
Advantec 301200 Stainless Steel Filter Holder, Syringe; 25mm dia WW-02928-20 Mfr # 301200
COMPARE
Stainless Steel 25 Female luer
₹61,500.00INR / Each
Advantec 304500 Stainless Steel Filter Holder, In-Line; 25mm dia WW-02929-10 Mfr # 304500
COMPARE
Stainless Steel 25 1/4\" NPT(F)
₹79,600.00INR / Each
Advantec 304700 Stainless Steel Filter Holder, In-Line; 47mm dia WW-02929-20 Mfr # 304700
COMPARE
Stainless Steel 47 1/4\" NPT(F)
₹155,000.00INR / Each