Advantec PFA Filter Holders
Use the PFA holders with chemically aggressive liquids
  • Maximum operating temperature is 250°F (121°C)
  • Autoclavable
Dual support screens allow for flow in either direction. Exterior locking ring designed for quick assembly without tearing the membrane. They are ideal for sterilizing and filtering ultra clean liquids under positive pressure and filtering small volumes of high-purity acids, alkalis, or organic solvents. Body and support screens of the filters are made of PFA; O-ring is made of perfluoroelastomer.
19,000.00 - ₹44,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Body Material Filter Diameter (mm)
Availability Pricing
Advantec 43307010 In-Line PFA Filter Holder, 25 mm; 1/Pk WW-06644-60 Mfr # 43307010
COMPARE
PFA 25
₹19,000.00INR / Each
Advantec 43307020 In-Line PFA Filter Holder, 47 mm; 1/Pk WW-06644-62 Mfr # 43307020
COMPARE
PFA 47
₹44,100.00INR / Each