Advantec Narrow Range pH Test Papers
Choose only the ranges you need, but keep the fine 0.2 to 0.4 pH unit resolution
These pH test paper booklets feature narrow pH unit graduation (0.2 to 0.4 pH) and can be used for severe pH measuring. Each leaflet is 1/4" x 2-3/4" (7 x 70-mm). There are 20 leaflets per booklet and 10 booklets per box.
2,900.00 - ₹6,000.00INR / Box of 1
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Measurement Points
Availability Pricing
Advantec Narrow Range pH Paper Booklet, Cresol red (0.4-2.0, 7.2-8.8 pH), 200/bx WW-59201-01 Mfr # 07010010
COMPARE
0.4 to 2.0 pH; 7.2 to 8.8 pH
₹2,900.00INR / Box of 1
Advantec Narrow Range pH Paper Booklet, Thymol blue (1.4-3, 8-9.6 pH), 200/bx WW-59201-03 Mfr # 07010020
COMPARE
1.4 to 3 pH; 8 to 9.6 pH
₹2,900.00INR / Box of 1
Advantec Narrow Range pH Paper Booklet, Bromophenol blue (2.8-4.4 pH), 200/bx WW-59201-05 Mfr # 07010030
COMPARE
2.8 to 4.4 pH
₹2,900.00INR / Box of 1
Advantec Narrow Range pH Paper Booklet, Bromocresol green (4.0-5.6 pH), 200/bx WW-59201-07 Mfr # 07010040
COMPARE
4.0 to 5.6 pH
₹6,000.00INR / Box of 1
Advantec 07010050 Narrow Range pH Paper Booklet, Methyl red (5.4-7.0 pH), 200/bx WW-59201-09 Mfr # 07010050
COMPARE
5.4 to 7 pH
₹2,900.00INR / Box of 1
Advantec Narrow Range pH Paper Booklet, Bromothymol blue (6.2-7.8 pH), 200/bx WW-59201-11 Mfr # 07010060
COMPARE
6.2 to 7.8 pH
₹2,900.00INR / Box of 1
Advantec Narrow Range pH Paper Booklet, Alizarin yellow (10.0-12.0 pH), 200/bx WW-59201-13 Mfr # 07010070
COMPARE
10 to 12 pH
₹2,900.00INR / Box of 1
Advantec 07010080 Narrow Range pH Paper Booklet, Alkali blue (11.0-13.6 pH), 200/bx WW-59201-15 Mfr # 07010080
COMPARE
11 to 13.6 pH
₹2,900.00INR / Box of 1
Advantec Narrow Range pH Paper Booklet, Phenol blue (3.2-5.6 pH), 200/bx WW-59201-17 Mfr # 07010090
COMPARE
3.2 to 5.6 pH
₹3,400.00INR / Box of 1
Advantec Narrow Range pH Paper Booklet, Chlorophenol red (5.0-6.6 pH), 200/bx WW-59201-19 Mfr # 07010100
COMPARE
5 to 6.6 pH
₹2,900.00INR / Box of 1
Advantec 07010110 Narrow Range pH Paper Booklet, MR/BTB (5.0-8.0 pH), 200/bx WW-59201-21 Mfr # 07010110
COMPARE
5 to 8.0 pH
₹2,900.00INR / Box of 1
Showing 1- 11 of 11