Advantec Narrow Range pH Paper Booklet, Chlorophenol red (5.0-6.6 pH), 200/bx
Representative image only.
Advantec Narrow Range pH Paper Booklet, Chlorophenol red (5.0-6.6 pH), 200/bx
Advantec | Mfr: 07010100| Item: 5920119
Choose only the ranges you need, but keep the fine 0.2 to 0.4 pH unit resolution
₹3,700.00INR / Box of 1
₹3,700.00INR / Box of 1
Product Description
More About this Item
These pH test paper booklets feature narrow pH unit graduation (0.2 to 0.4 pH) and can be used for severe pH measuring. Each leaflet is 1/4" x 2-3/4" (7 x 70-mm). There are 20 leaflets per booklet and 10 booklets per box.
Specifications
  • TestpH
  • Measurement Points5 to 6.6 pH
  • Tests Per Kit200
  • DescriptionNarrow Range pH Paper Booklet, Chlorophenol red (5.0-6.6 pH), 200/bx