Advantec Grade QR100 Quartz Fiber Filters
For analytical or gravimetric studies
Quartz fiber filters have very low trace metal content and can withstand temperatures up to 1832°F (1000°C). Use to sample acidic gases at high temperatures or for air pollution analysis. The QR100 Grade filters offer superior chemical resistance and will not absorb acidic gases.
22,300.00 - ₹82,400.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm)
Availability Pricing
Advantec QR10037MM Grade QR100 Quartz Fiber Filters, 37-mm dia; 100/Box 0664950 Mfr # QR10037MM
COMPARE
37
₹33,800.00INR / Pkg of 100
Advantec QR10050MM Grade QR100 Quartz Fiber Filters, 50-mm dia; 100/Box 0664954 Mfr # QR10050MM
COMPARE
50
₹46,100.00INR / Pkg of 100
Advantec QR10090MM QR100 Quartz Fiber Filters, 82 mm Dia; 100/Box 0664956 Mfr # QR10082MM
COMPARE
82
₹82,400.00INR / Pkg of 100
Advantec QR10025MM Grade QR100 Quartz Fiber Filters, 25-mm dia; box of 100. 0665820 Mfr # QR10025MM
COMPARE
25
₹22,300.00INR / Pkg of 100