Advantec QR10050MM Grade QR100 Quartz Fiber Filters, 50-mm dia; 100/Box
Representative image only.
Advantec QR10050MM Grade QR100 Quartz Fiber Filters, 50-mm dia; 100/Box
Advantec | Mfr: QR10050MM| Item: 0664954
For analytical or gravimetric studies
₹46,100.00INR / Pkg of 100
₹46,100.00INR / Pkg of 100
Product Description
Key Features
  • 1 year warranty
More About this Item
Quartz fiber filters have very low trace metal content and can withstand temperatures up to 1832°F (1000°C). Use to sample acidic gases at high temperatures or for air pollution analysis. The QR100 Grade filters offer superior chemical resistance and will not absorb acidic gases.
1 Year Warranty
Specifications
  • MaterialQuartz Microfiber
  • Diameter (mm)50
  • Diameter (cm)5
  • DescriptionGrade QR100 Quartz Fiber Filters, 50-mm dia; 100/Box