Advantec Glass and Quartz Fiber Filters
For analytical or gravimetric studies
  • Borosilicate glass filters create a biologically inert filter matrix that is resistant to both chemicals and pH extremes
Use as prefilters to extend membrane life, for water/air pollution analysis, or liquid clarification. Filters without binder withstand up to 932°F (500°C); filters with binder withstand up to 250°F (120°C). Grades GC90, GS25, and DP70 with binder offer higher wet strength and lower fiber release. Quartz fiber filters have very low trace metal content and can withstand up to 1832°F (1000°C). Choose quartz fiber filters to sample acidic gases at high temperatures or for air pollution analysis.
2,100.00 - ₹47,720.00INR / Pkg of 100
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (micron)
Availability Pricing
Advantec GA200110MM GA200 Glass Fiber Filters, 1.0 um, 110 mm Dia; 50/Box WW-06646-55 Mfr # GA200110MM
COMPARE
110 Glass Microfiber 1
₹19,270.00INR / Pkg of 50
Advantec GA200150MM GA200 Glass Fiber Filters, 1.0 um, 150 mm Dia; 50/Box WW-06646-62 Mfr # GA200150MM
COMPARE
150 Glass Microfiber 1
₹36,080.00INR / Pkg of 50
Advantec GA20070MM GA200 Glass Fiber Filters, 1.0 um, 70 mm Dia; 50/Box WW-06646-64 Mfr # GA20070MM
COMPARE
70 Glass Microfiber 1
₹11,180.00INR / Pkg of 50
Advantec GA5524MM GA55 Glass Fiber Filters, 1.6 um, 24 mm Dia; 100/Box WW-06646-72 Mfr # GA5524MM
COMPARE
24 Glass Microfiber 0.6
₹3,580.00INR / Pkg of 100
Advantec GA5537MM GA55 Glass Fiber Filters, 1.6 um, 37 mm Dia; 100/Box WW-06646-73 Mfr # GA5537MM
COMPARE
37 Glass Microfiber 0.6
₹4,500.00INR / Pkg of 100
Advantec GA558X10IN GA55 Glass Fiber Filters, 1.6 um, 8" x 10"; 100/Box WW-06646-74 Mfr # GA558X10IN
COMPARE
Glass Microfiber 0.6
₹37,200.00INR / Pkg of 100
Advantec GB14042.5MM GB140 Glass Fiber Filters, 0.4 um, 42.5 mm Dia; 100/Box WW-06646-85 Mfr # GB14042.5MM
COMPARE
42.5 Glass Microfiber 0.4
₹6,600.00INR / Pkg of 100
Advantec GC50110MM GC50 Glass Fiber Filters, 0.5 um, 110 mm Dia; 100/Box WW-06646-93 Mfr # GC50110MM
COMPARE
110 Glass Microfiber 0.5
₹22,040.00INR / Pkg of 100
Advantec GC50150MM GC50 Glass Fiber Filters, 0.5 um, 150 mm Dia; 100/Box WW-06646-94 Mfr # GC50150MM
COMPARE
150 Glass Microfiber 0.6
₹46,660.00INR / Pkg of 100
Advantec GC5021MM GC50 Glass Fiber Filters, 0.5 um, 21 mm Dia; 100/Box WW-06646-95 Mfr # GC5021MM
COMPARE
21 Glass Microfiber 0.7
₹5,240.00INR / Pkg of 100
Advantec GC50257MM GC50 Glass Fiber Filters, 0.5 um, 257 mm Dia; 50/Box WW-06647-94 Mfr # GC50257MM
COMPARE
257 Glass Microfiber 0.1
₹37,700.00INR / Pkg of 100
Advantec GC5070MM GC50 Glass Fiber Filters, 0.5 um, 70 mm Dia; 100/Box WW-06656-02 Mfr # GC5070MM
COMPARE
70 Glass Microfiber 0.5
₹11,700.00INR / Pkg of 100
Advantec GC5075MM GC50 Glass Fiber Filters, 0.5 um, 75 mm Dia; 100/Box WW-06656-03 Mfr # GC5075MM
COMPARE
75 Glass Microfiber 0.5
₹14,850.00INR / Pkg of 100
Advantec GC5082MM GC50 Glass Fiber Filters, 0.5 um, 82 mm Dia; 100/Box WW-06656-04 Mfr # GC5082MM
COMPARE
82 Glass Microfiber 0.5
₹17,800.00INR / Pkg of 100
Advantec GD12042.5MM GD120 Glass Fiber Filters, 0.9 um, 42.5 mm Dia; 50/Box WW-06656-15 Mfr # GD12042.5MM
COMPARE
42.5 Glass Microfiber 0.9
₹2,100.00INR / Pkg of 50
Advantec GF7513MM GF75 Glass Fiber Filters, 0.3 um, 13 mm Dia; 100/Box WW-06656-22 Mfr # GF7513MM
COMPARE
13 Glass Microfiber 0.33
₹7,700.00INR / Pkg of 100
Advantec GF7537MM GF75 Glass Fiber Filters, 0.3 um, 37 mm Dia; 100/Box WW-06656-32 Mfr # GF7537MM
COMPARE
37 Glass Microfiber 0.3
₹13,310.00INR / Pkg of 100
Advantec GB140110MM Glass fiber filter, GB140, 110-mm dia; box of 100 WW-06657-15 Mfr # GB140110MM
COMPARE
110 Glass Microfiber
₹25,730.00INR / Pkg of 100
Advantec GF7542.5MM Glass fiber filter, GF75, 42.5-mm dia; box of 100 WW-06657-41 Mfr # GF7542.5MM
COMPARE
42.5 Glass Microfiber
₹14,990.00INR / Pkg of 100
Advantec GF75110MM Glass fiber filter, GF75, 110-mm dia; box of 100 WW-06657-45 Mfr # GF75110MM
COMPARE
110 Glass Microfiber
₹47,720.00INR / Pkg of 50
Showing 1 - 20 of 21