Advantec GF75110MM Glass fiber filter, GF75, 110-mm dia; box of 100
Representative image only.
Advantec GF75110MM Glass fiber filter, GF75, 110-mm dia; box of 100
Advantec | Mfr: GF75110MM| Item: 0665745
For analytical or gravimetric studies
₹74,000.00INR / Pkg of 50
₹74,000.00INR / Pkg of 50
Product Description
Key Features
  • Borosilicate glass filters create a biologically inert filter matrix that is resistant to both chemicals and pH extremes
  • 1 year warranty
More About this Item
Use as prefilters to extend membrane life, for water/air pollution analysis, or liquid clarification. Filters without binder withstand up to 932°F (500°C); filters with binder withstand up to 250°F (120°C). Grades GC90, GS25, and DP70 with binder offer higher wet strength and lower fiber release. Quartz fiber filters have very low trace metal content and can withstand up to 1832°F (1000°C). Choose quartz fiber filters to sample acidic gases at high temperatures or for air pollution analysis.
1 Year Warranty
Specifications
  • MaterialGlass Microfiber
  • Diameter (mm)110
  • Diameter (cm)11
  • DescriptionGlass fiber filter, GF75, 110-mm dia; box of 100