Advantec Disposable Ultrafiltration Units
Concentrate, separate, and purify your biological samples without a centrifuge
  • Three molecular weight cutoffs
  • 40 times maximum concentration
These disposable ultrafiltration units are designed for ease in concentrating, separating, or otherwise purifying small volumes (<2 mL) of liquid samples, on the basis of molecular weight cutoff using positive pressure. The unit is constructed of polypropylene cap, acrylic retentive cell, and filtrate cell. Filter media is polysulfone. To use, simply dispense sample into the retentive cell by pipette, place cap on retentive cell, and inject 10 mL of air with a syringe. Remove cap to stop filtration. Sterilization: 25% ethanol, 5% formalin.
25,570.00INR / Pkg of 24
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Nominal Molecular Weight Limit (Mwl) (Dalton)
Availability Pricing
Advantec USY-1 Disposable Ultrafiltration Unit, 10K MWCO, pack of 24 WW-02910-50 Mfr # USY-1
COMPARE
10000
₹25,570.00INR / Pkg of 24
Advantec USY-5 Disposable Ultrafiltration Unit, 50K MWCO, pack of 24 WW-02910-52 Mfr # USY-5
COMPARE
50000
₹25,570.00INR / Pkg of 24
Advantec USY-20 Disposable Ultrafiltration Unit, 200K MWCO, pack of 24 WW-02910-54 Mfr # USY-20
COMPARE
200000
₹25,570.00INR / Pkg of 24