Advantec USY-1 Disposable Ultrafiltration Unit, 10K MWCO, pack of 24

Representative image only.

Advantec USY-1 Disposable Ultrafiltration Unit, 10K MWCO, pack of 24

Advantec | Mfr: USY-1| Item: 0291050

Concentrate, separate, and purify your biological samples without a centrifuge

₹39,200.00INR / Pkg of 24

₹39,200.00INR / Pkg of 24
Product Description

Includes

24 retentive cells, 24 filtrate cells, and 4 reusable caps.

Key Features

  • Three molecular weight cutoffs
  • 40 times maximum concentration
  • 1 year warranty

More About this Item

These disposable ultrafiltration units are designed for ease in concentrating, separating, or otherwise purifying small volumes (<2 mL) of liquid samples, on the basis of molecular weight cutoff using positive pressure. The unit is constructed of polypropylene cap, acrylic retentive cell, and filtrate cell. Filter media is polysulfone. To use, simply dispense sample into the retentive cell by pipette, place cap on retentive cell, and inject 10 mL of air with a syringe. Remove cap to stop filtration. Sterilization: 25% ethanol, 5% formalin.

1 Year Warranty
Specifications
  • Hold-Up Volume (mL)0.02
  • Max Pressure (PSI)43
  • Max Pressure (bar)2.9
  • Max Temperature (° C)50
  • Max Temperature (° F)122
  • Nominal Molecular Weight Limit (Mwl) (Dalton)10000
  • Filtration Area (cm²)2.0
  • Maxi Pressure (Mpa)0.29
  • Max Molecular Weight Cut Off (Mwco) (Dalton)10000
  • Min Volume Range (mL)0.02
  • Max Volume Range (mL)2
  • DescriptionDisposable Ultrafiltration Unit, 10K MWCO, pack of 24
  • Warranty1 year
Reviews
Q&A