Advantec Air Sampling Filters, PTFE Coated, Hydrophobic
Composite filters ideal for sampling airborne particulates
Composite Filters are specially developed for the measurement of dusts and particulate matter in the air. These PTFE coated filters are naturally hydrophobic and unaffected by humidity. Thermo-stable to 500°F (260°C).
50,900.00 - ₹69,800.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm)
Availability Pricing
Advantec PG6025MM Air Sampling Filters, Composite, 25-mm dia disc; pack of 100. 0664960 Mfr # PG6025MM
COMPARE
25
₹50,900.00INR / Pkg of 100
Advantec PG6047MM Air Sampling Filters, Composite, 47-mm dia disc; pack of 100. 0664961 Mfr # PG6047MM
COMPARE
47
₹69,800.00INR / Pkg of 100