Advantec PG6047MM Air Sampling Filters, Composite, 47-mm dia disc; pack of 100.
Representative image only.
Advantec PG6047MM Air Sampling Filters, Composite, 47-mm dia disc; pack of 100.
Advantec | Mfr: PG6047MM| Item: 0664961
Composite filters ideal for sampling airborne particulates
₹69,800.00INR / Pkg of 100
₹69,800.00INR / Pkg of 100
Product Description
Key Features
  • 1 year warranty
More About this Item
Composite Filters are specially developed for the measurement of dusts and particulate matter in the air. These PTFE coated filters are naturally hydrophobic and unaffected by humidity. Thermo-stable to 500°F (260°C).
1 Year Warranty
Specifications
  • MaterialPTFE
  • Diameter (mm)47
  • Diameter (cm)4.7
  • DescriptionAir Sampling Filters, Composite, 47-mm dia disc; pack of 100.