Advantec Air Sampling Filters, Glass Fiber
Glass fiber filters ideal for sampling airborne particulates
Glass Filters are binder-free and have a low trace metals content. Glass fibers are unaffected by humidity, highly resistant to chemical attack, and biologically inert. Used for high-volume sampling of aerosols, particulates, and metal contaminants.
10,300.00 - ₹52,200.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Advantec GB100R47MM Air Sampling Filters, Glass Fiber, 47-mm dia disc; pack of 100 0664675 Mfr # GB100R47MM
COMPARE
Air Sampling Filters, Glass Fiber, 47-mm dia disc; pack of 100
₹10,300.00INR / Pkg of 100
Advantec GB100R8X10IN GB100R Glass Fiber Air Sampling Filters, 0.6um, 8" x 10"; 50/Box 0664682 Mfr # GB100R8X10IN
COMPARE
GB100R Glass Fiber Air Sampling Filters, 0.6um, 8" x 10"; 50/Box
₹52,200.00INR / Pkg of 50