Advantec GB100R47MM Air Sampling Filters, Glass Fiber, 47-mm dia disc; pack of 100
Representative image only.
Advantec GB100R47MM Air Sampling Filters, Glass Fiber, 47-mm dia disc; pack of 100
Advantec | Mfr: GB100R47MM| Item: 0664675
Glass fiber filters ideal for sampling airborne particulates
₹10,300.00INR / Pkg of 100
₹10,300.00INR / Pkg of 100
Product Description
Key Features
  • 1 year warranty
More About this Item
Glass Filters are binder-free and have a low trace metals content. Glass fibers are unaffected by humidity, highly resistant to chemical attack, and biologically inert. Used for high-volume sampling of aerosols, particulates, and metal contaminants.
1 Year Warranty
Specifications
  • MaterialGlass Microfiber
  • Diameter (mm)47
  • Diameter (cm)4.7
  • DescriptionAir Sampling Filters, Glass Fiber, 47-mm dia disc; pack of 100