Adhesive Entrance Mats
Mat frame system can be used at lab or office entrance to keep work areas clean
Entrance mat system is designed to remove dust, dirt, and other particulate from worker's shoes, carts, and dollies before being tracked into labs or office space. Mats are available in 30 or 60 layer versions and in white or blue. Each layer of the mat is fully coated with a particle-grabbing adhesive and an antimicrobial agent, and is constructed of 1.8-mil (50-micron) low-density polyethylene film. A printed number in the corner of each sheet indicates how many layers remain, and ensures at least one layer remains before replacement. Frames are all white and feature a polyester web-lock pad on the bottom to prevent it from slipping or sliding; simply replace with refill mats when needed.
17,700.00 - ₹47,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Color Description
Availability Pricing
Purus PFM253131W01 Adhesive Mat Starter Kit, white, 30 layer 6410030 Mfr # PFM253131W01
COMPARE
White Adhesive Mat Starter Kit, white, 30 layer
₹18,100.00INR / Each
Purus PFM253131W02 Adhesive Mat Starter Kit, blue, 30 layer 6410035 Mfr # PFM253131W02
COMPARE
Blue Adhesive Mat Starter Kit, blue, 30 layer
₹17,700.00INR / Each
Purus PF2430-34W Adhesive Mat Refill, white, 30 layer, 4/pk 6410036 Mfr # PF2430-34W
COMPARE
White Adhesive Mat Refill, white, 30 layer, 4/pk
₹19,700.00INR / Pkg of 4
Purus PF2430-34B Adhesive Mat Refill, blue, 30 layer, 4/pk 6410037 Mfr # PF2430-34B
COMPARE
Blue Adhesive Mat Refill, blue, 30 layer, 4/pk
₹19,700.00INR / Pkg of 4
Purus PFM253161W01 Adhesive Mat Starter Kit, white, 60 layer 6410040 Mfr # PFM253161W01
COMPARE
White Adhesive Mat Starter Kit, white, 60 layer
₹23,400.00INR / Each
Purus PFM253161W02 Adhesive Mat Starter Kit, blue, 60 layer 6410045 Mfr # PFM253161W02
COMPARE
Blue Adhesive Mat Starter Kit, blue, 60 layer
₹23,400.00INR / Each
Purus PF2430-64W Adhesive Mat Refill, white, 60 layer, 4/pk 6410046 Mfr # PF2430-64W
COMPARE
White Adhesive Mat Refill, white, 60 layer, 4/pk
₹37,200.00INR / Pkg of 4
Purus PF2430-64B Adhesive Mat Refill, blue, 60 layer, 4/pk 6410047 Mfr # PF2430-64B
COMPARE
Blue Adhesive Mat Refill, blue, 60 layer, 4/pk
₹47,000.00INR / Pkg of 4
Showing 1- 8 of 8