Purus PFM253131W02 Adhesive Mat Starter Kit, blue, 30 layer
Representative image only.
Purus PFM253131W02 Adhesive Mat Starter Kit, blue, 30 layer
Purus | Mfr: PFM253131W02| Item: 6410035
Mat frame system can be used at lab or office entrance to keep work areas clean
₹17,700.00INR / Each
₹17,700.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Entrance mat system is designed to remove dust, dirt, and other particulate from worker's shoes, carts, and dollies before being tracked into labs or office space. Mats are available in 30 or 60 layer versions and in white or blue. Each layer of the mat is fully coated with a particle-grabbing adhesive and an antimicrobial agent, and is constructed of 1.8-mil (50-micron) low-density polyethylene film. A printed number in the corner of each sheet indicates how many layers remain, and ensures at least one layer remains before replacement. Frames are all white and feature a polyester web-lock pad on the bottom to prevent it from slipping or sliding; simply replace with refill mats when needed.
1 Year Warranty
Specifications
  • MaterialLDPE
  • Length (in)30
  • Length (cm)76.2
  • ColorBlue
  • DescriptionAdhesive Mat Starter Kit, blue, 30 layer