Add-On Accessories, 304 Stainless Steel
These accessories include the necessary snap-in support pegs to quickly and easily attach to the rack. Drain basket holds small items such as stir bars, stoppers, and pipette tips. Drain shelf is ideal for drying and storing various glassware items. Pipette rack hangs at an angle to allow for complete drainage of pipettes. Funnel rack holds four funnels, volumetric flasks, or small cylinders. Cylinder yoke holds two cylinders. Flask holder accepts one large flask, cylinder, or separatory funnel. Glove dispenser provides a convenient spot for one box of gloves. Screen inserts fit into the drip trough and increase the drying capacity of the rack.
10,400.00 - ₹22,200.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Drain basket for modular stainless steel drying racks
₹10,400.00INR / Each
COMPARE
Drain shelf for modular stainless steel drying racks
₹15,700.00INR / Each
COMPARE
Pipette rack for modular stainless steel drying racks
₹15,700.00INR / Each
COMPARE
Funnel rack for modular stainless steel drying racks
₹15,700.00INR / Each
COMPARE
Cylinder yoke for modular stainless steel drying racks
₹15,700.00INR / Each
COMPARE
Flask holder for modular stainless steel drying racks
₹10,400.00INR / Each
COMPARE
Screen insert for 32-peg modular stainless steel drying racks
₹20,000.00INR / Each
COMPARE
Screen insert for 50-peg modular stainless steel drying racks
₹22,200.00INR / Each
COMPARE
Screen insert for 60-peg modular stainless steel drying racks
₹19,400.00INR / Each
Showing 1- 9 of 9