Funnel rack for modular stainless steel drying racks
Representative image only.
Funnel rack for modular stainless steel drying racks
| Item: 6703573
₹18,000.00INR / Each
₹18,000.00INR / Each
Buy It Together
Current Item
Funnel rack for modular stainless steel drying racks
Funnel rack for modular stainless steel drying racks
6703573
₹18,000.00INR / Each
+
Modular stainless steel drying rack, 32 black pegs
6703504
₹171,000.00INR / Each
=
Total Price 189000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These accessories include the necessary snap-in support pegs to quickly and easily attach to the rack. Drain basket holds small items such as stir bars, stoppers, and pipette tips. Drain shelf is ideal for drying and storing various glassware items. Pipette rack hangs at an angle to allow for complete drainage of pipettes. Funnel rack holds four funnels, volumetric flasks, or small cylinders. Cylinder yoke holds two cylinders. Flask holder accepts one large flask, cylinder, or separatory funnel. Glove dispenser provides a convenient spot for one box of gloves. Screen inserts fit into the drip trough and increase the drying capacity of the rack.
Lifetime Warranty
Specifications
  • Width (in)14
  • Width (mm)356
  • DescriptionFunnel rack for modular stainless steel drying racks