Adam Equipment Polaris Precision Toploading Balances
View step-by-step instructions, fast results, and real-time application status on a large multiline, backlit display
  • Built-in USB, RS-232, Ethernet, and Bluetooth® connections for seamless data transfer
  • Store up to 200 ingredients and 240,000 test data records in the internal memory
  • Color-changing backlight indicates under, over, or within acceptable limits in checkweighing mode
  • GLP-compliant reporting includes time, date, balance number, and a verification of the calibration
  • View display in English, German, French, Spanish, Portuguese, or Italian
  • Passcode security function prevents unintentional menu changes
  • Sealed keypad protects against dirt and spills
  • ABS housing and 304 SS pan offer excellent durability
The Polaris precision toploading balances deliver high weighing accuracy and repeatability at an affordable price. A 7” LCD display provides intuitive, easy-to-follow instructions and shows fast results and application status at a glance. Advanced connectivity options include two USBs, bidirectional RS-232, Ethernet, and Bluetooth. Connect a computer, printer, or flash drive for a seamless data transfer.
Using the checkweighing function is made easier with color-changing backlight, an audible alarm, and prompt box display to indicate if the weight is under, over, or within acceptable limits. A multi-level security feature allows one administrator and up to 99 operators, ensuring balance use only by authorized users.
429,000.00 - ₹968,000.00INR / Each
22 variations of this product are available.
See All Product Options Below
22 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Adam Equipment PPB 223i Polaris Precision Toploading Balance, 220g x 0.001g with LED Illuminated Weighing Chamber and Internal Calibration 2060576 Mfr # PPB 223i
COMPARE
Internal 220 0.001
₹552,000.00INR / Each
Adam Equipment PPB 423i Polaris Precision Toploading Balance, 420g x 0.001g with LED Illuminated Weighing Chamber and Internal Calibration 2060577 Mfr # PPB 423i
COMPARE
Internal 420 0.001
₹636,000.00INR / Each
Adam Equipment PPB 623i Polaris Precision Toploading Balance, 620g x 0.001g with LED Illuminated Weighing Chamber and Internal Calibration 2060578 Mfr # PPB 623i
COMPARE
Internal 620 0.001
₹719,000.00INR / Each
Adam Equipment PPB 823i Polaris Precision Toploading Balance, 820g x 0.001g with LED Illuminated Weighing Chamber and Internal Calibration 2060579 Mfr # PPB 823i
COMPARE
Internal 820 0.001
₹802,000.00INR / Each
Adam Equipment PPB 1223i Polaris Precision Toploading Balance, 1220g x 0.001g with LED Illuminated Weighing Chamber and Internal Calibration 2060580 Mfr # PPB 1223i
COMPARE
Internal 1220 0.001
₹885,000.00INR / Each
Adam Equipment PPB 1623i Polaris Precision Toploading Balance, 1620g x 0.001g with LED Illuminated Weighing Chamber and Internal Calibration 2060581 Mfr # PPB 1623i
COMPARE
Internal 1620 0.001
₹968,000.00INR / Each
Adam Equipment PPB 223e Polaris Precision Toploading Balance, 220g x 0.001g with LED Illuminated Weighing Chamber and External Calibration 2060582 Mfr # PPB 223e
COMPARE
External 220 0.001
₹461,000.00INR / Each
Adam Equipment PPB 423e Polaris Precision Toploading Balance, 420g x 0.001g with LED Illuminated Weighing Chamber and External Calibration 2060583 Mfr # PPB 423e
COMPARE
External 420 0.001
₹530,000.00INR / Each
Adam Equipment PPB 623e Polaris Precision Toploading Balance, 620g x 0.001g with LED Illuminated Weighing Chamber and External Calibration 2060584 Mfr # PPB 623e
COMPARE
External 620 0.001
₹600,000.00INR / Each
Adam Equipment PPB 823e Polaris Precision Toploading Balance, 820g x 0.001g with LED Illuminated Weighing Chamber and External Calibration 2060585 Mfr # PPB 823e
COMPARE
External 820 0.001
₹669,000.00INR / Each
Adam Equipment PPB 1223e Polaris Precision Toploading Balance, 1220g x 0.001g with LED Illuminated Weighing Chamber and External Calibration 2060586 Mfr # PPB 1223e
COMPARE
1220 0.001
₹738,000.00INR / Each
Adam Equipment PPB 1623e Polaris Precision Toploading Balance, 1620g x 0.001g with LED Illuminated Weighing Chamber and External Calibration 2060587 Mfr # PPB 1623e
COMPARE
1620 0.001
₹807,000.00INR / Each
Adam Equipment PTB 2602i Polaris Precision Toploading Balance, 2600g x 0.01g with Internal Calibration 2060588 Mfr # PTB 2602i
COMPARE
Internal 2600 0.01
₹552,000.00INR / Each
Adam Equipment PTB 4602i Polaris Precision Toploading Balance, 4600g x 0.01g with Internal Calibration 2060589 Mfr # PTB 4602i
COMPARE
4600 0.01
₹636,000.00INR / Each
Adam Equipment PTB 6202i Polaris Precision Toploading Balance, 6200g x 0.01g with internal calibration 2060590 Mfr # PTB 6202i
COMPARE
6200 0.01
₹719,000.00INR / Each
Adam Equipment PTB 2602e Polaris Precision Toploading Balance, 2600g x 0.01 with External Calibration 2060591 Mfr # PTB 2602e
COMPARE
2600 0.01
₹622,000.00INR / Each
Adam Equipment PTB 4602e Polaris Precision Toploading Balance, 4600g x 0.01g with External Calibration 2060592 Mfr # PTB 4602e
COMPARE
4600 0.01
₹706,000.00INR / Each
Adam Equipment PTB 6202e Polaris Precision Toploading Balance, 6200g x 0.01g with External Calibration 2060593 Mfr # PTB 6202e
COMPARE
External 6200 0.01
₹789,000.00INR / Each
Adam Equipment PTB 8202e Polaris Precision Toploading Balance, 8200g x 0.01g with External Calibration 2060594 Mfr # PTB 8202e
COMPARE
External 8200 0.01
₹872,000.00INR / Each
Adam Equipment PBB 6001e Polaris Precision Toploading Balance, 6000g x 0.1g with External Calibration 2060595 Mfr # PBB 6001e
COMPARE
External 6000 0.1
₹429,000.00INR / Each
Showing 1 - 20 of 22