Adam Equipment Polaris Analytical Balances
Brightly lit weighing chamber provides clear visibility of samples
  • Intuitive, easy-to-use multiline display
  • Built-in USB, RS-232, Ethernet, and Bluetooth® connections for seamless data transfer
  • Store up to 200 ingredients and 240,000 test data records in the internal memory
  • Color-changing backlight indicates under, over, or within acceptable limits in checkweighing mode
  • GLP-compliant reporting includes time, date, balance number, and a verification of the calibration
  • View display in English, German, French, Spanish, Portuguese, or Italian
  • Passcode security function prevents unintentional menu changes
  • Sealed keypad protects against dirt and spills
  • ABS housing and 304 SS pan offer excellent durability
The Polaris analytical balances deliver high weighing accuracy and repeatability at an affordable price. A bright, LED light in the draft shield allows clear visibility of samples on the weighing pan. View step-by-step instructions, fast results, and real-time application status on a 7.4” multiline, backlit display. Advanced connectivity options include two USBs, bidirectional RS-232, Ethernet, and Bluetooth. Connect a computer, printer, or flash drive for a seamless data transfer.
Using the checkweighing function is made easier with color-changing backlight, an audible alarm, and prompt box display to indicate if the weight is under, over, or within acceptable limits. A multi-level security feature allows one administrator and up to 99 operators, ensuring balance use only by authorized users.
747,000.00 - ₹1,190,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g)
Availability Pricing
Adam Equipment PAB 124i Polaris Analytical Balance, 120g x 0.0001g with LED Illuminated Weighing Chamber and Internal Calibration 2060568 Mfr # PAB 124i
COMPARE
Internal 120
₹830,000.00INR / Each
Adam Equipment PAB 224i Polaris Analytical Balance, 220g x 0.0001g with LED Illuminated Weighing Chamber and Internal Calibration 2060569 Mfr # PAB 224i
COMPARE
Internal 220
₹927,000.00INR / Each
Adam Equipment PAB 324i Polaris Analytical Balance, 320g x 0.0001g with LED Illuminated Weighing Chamber and Internal Calibration 2060570 Mfr # PAB 324i
COMPARE
Internal 320
₹1,024,000.00INR / Each
Adam Equipment PAB 424i Polaris Analytical Balance, 420g x 0.0001g with LED Illuminated Weighing Chamber and Internal Calibration 2060571 Mfr # PAB 424i
COMPARE
Internal 420
₹1,190,000.00INR / Each
Adam Equipment PAB 124e Polaris Analytical Balance, 120g x 0.0001g with LED Illuminated Weighing Chamber and External Calibration 2060572 Mfr # PAB 124e
COMPARE
External 120
₹747,000.00INR / Each
Adam Equipment PAB 224e Polaris Analytical Balance, 220g x 0.0001g with LED Illuminated Weighing Chamber and External Calibration 2060573 Mfr # PAB 224e
COMPARE
External 220
₹844,000.00INR / Each
Adam Equipment PAB 324e Polaris Analytical Balance, 320g x 0.0001g with LED Illuminated Weighing Chamber and External Calibration 2060574 Mfr # PAB 324e
COMPARE
External 320
₹941,000.00INR / Each
Adam Equipment PAB 424e Polaris Analytical Balance, 420g x 0.0001g with LED Illuminated Weighing Chamber and External Calibration 2060575 Mfr # PAB 424e
COMPARE
External 420
₹1,107,000.00INR / Each
Showing 1- 8 of 8