Adam Equipment LHS Crane Scales
Flexible crane scales make it easy to measure suspended loads
  • Three-button infrared remote control
  • Color-coded keypad
  • Three weighing units (kg, lb, Newtons)
  • Display hold function
  • 360-degree rotating display
  • 360-degree swivel shackle and bolt
  • External calibration
  • Polycarbonate indicator housing (SHS only)
  • Cast aluminum magnesium indicator housing (LHS only)
  • Chrome-plated steel hook with safety catch
  • Automatic power-off
  • Low-battery indicator
  • Rechargeable battery
Designed with safety in mind, each scale features a standard remote control that allows operation at a safe distance. The rotating hook with safety catch ensures stability and keeps the suspended item securely in place. High-contrast red LED display and indicators are easy to read in any lighting conditions. A handy hold function keeps the reading visible on the display after the weight is removed, allowing the operator to safely record the weight.
Built-in rechargeable battery - Alows the scale to be used in areas where power is not available.
Simple, Flexible Operation - Use the included remote control or the colourcoded keypad to safely operate the scale’s functions. The bright-yellow tare/zero key is easy to spot for quick recognition.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Adam Equipment LHS 1000A Rechargeable Battery Crane Scales; 500kg x 0.1kg 120 V 1171012 Mfr # LHS 1000A
COMPARE
500 1000 120
Call For Price
Adam Equipment LHS 3000A Rechargeable Battery Crane Scales; 1500kg x 0.2kg 120 V 1171013 Mfr # LHS 3000A
COMPARE
1500 3000 100 to 240
Call For Price
Adam Equipment LHS 4000A Rechargeable Battery Crane Scales; 2000kg x 0.5kg 120 V 1171014 Mfr # LHS 4000A
COMPARE
2000 4000 100 to 240
Call For Price
Adam Equipment LHS 1000A (220V) Rechargeable Battery Crane Scales; 500kg x 0.1kg, 220V 1171042 Mfr # LHS 500
COMPARE
500 1103.8 100 to 240
Call For Price
Adam Equipment LHS 3000A (220V) Rechargeable Battery Crane Scales; 1500kg x 0.2kg, 220V 1171043 Mfr # LHS 1500
COMPARE
1500 3311.3 230
Call For Price
Adam Equipment LHS 4000A Rechargeable Battery Crane Scales; 2000kg x 0.5kg, 220V 1171044 Mfr # LHS 2000
COMPARE
2000 4415 230