Adam Equipment Cruiser® CDT Bench Counting Scales
Automatically refines piece weight for greater accuracy
  • Ideal for fast, accurate bulk counting
  • View weight, unit weight, and count all at once on three backlit LCDs
  • Color-coded keys provide quick recognition of often used buttons
  • Store up to 100 items for frequently used parts
  • Internal counting resolution of 1:600,000 ensures precise results
  • RS-232 and optional USB outputs for communicating with a computer and printer
  • Stainless steel pan and solid ABS housing offers excellent durability
  • External calibration feature ensures accurate measurements
  • Internal rechargeable battery allows operation almost anywhere
Quickly and accurately count items with Cruiser® CDT dual counting scales. Scales can be used as a stand-alone unit or with a remote platform that can handle bulk parts with larger capacity. Scales without remote platform can be easily connected to a platform at a later time.
Weighing modes include parts counting, check counting, and net total weighing. Preset tare function and count preset (or check weighing) feature help speed repetitive weighing process. Use the product look up (PLU) feature to store descriptions, unit weights, and tare weights for up to 100 commonly used items. Simply enter the PLU number for the item to recall the data. Send data to a printer or computer using the RS-232 interface; optional USB interface simplifies data communication.
130,000.00 - ₹227,000.00INR / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Description
Availability Pricing
Adam Equipment CDT 16 Cruiser® Dual Counting Scale, 35lb / 16kg x 0.001lb / 0.5g WW-55001-00 Mfr # CDT 16
COMPARE
16 35 Counting Scale, 35lb / 16kg x 0.001lb / 0.5g
₹130,000.00INR / Each
Adam Equipment CDT 4 Cruiser® Dual Counting Scale, 8lb / 4kg x 0.0002lb / 0.1g WW-55001-01 Mfr # CDT 4
COMPARE
4 8 Counting Scale, 8lb / 4kg x 0.0002lb / 0.1g
₹130,000.00INR / Each
Adam Equipment CDT 8 Cruiser® Dual Counting Scale, 16lb / 8kg x 0.0005lb / 0.2g WW-55001-02 Mfr # CDT 8
COMPARE
8 16 Counting Scale, 16lb / 8kg x 0.0005lb / 0.2g
₹130,000.00INR / Each
Adam Equipment CDT 32 Cruiser® Dual Counting Scale, 70lb / 32kg x 0.002lb / 1g WW-55001-03 Mfr # CDT 32
COMPARE
32 70 Counting Scale, 70lb / 32kg x 0.002lb / 1g
₹130,000.00INR / Each
Adam Equipment CDT 48 Cruiser® Dual Counting Scale, 100lb / 48kg x 0.005lb / 2g WW-55001-04 Mfr # CDT 48
COMPARE
48 100 Counting Scale, 100lb / 48kg x 0.005lb / 2g
₹130,000.00INR / Each
Adam Equipment CDT 16-150 Cruiser® Dual Counting Scale, 35lb / 16kg with 330lb / 150kg Remote Platform WW-55001-05 Mfr # CDT 16-150
COMPARE
16 35 Counting Scale with Remote Platform 35lb / 16kg and 330lb / 150kg
₹189,000.00INR / Each
Adam Equipment CDT 16-60 Cruiser® Dual Counting Scale, 35lb / 16kg with 130lb / 60kg Remote Platform WW-55001-06 Mfr # CDT 16-60
COMPARE
16 35 Counting Scale with Remote Platform 35lb / 16kg and 130lb / 60kg
₹171,000.00INR / Each
Adam Equipment CDT 16-600 Cruiser® Dual Counting Scale, 35lb / 16kg with 1320lb / 600kg Remote Platform WW-55001-07 Mfr # CDT 16-600
COMPARE
16 35 Counting Scale with Remote Platform 35lb / 16kg and 1320lb / 600kg
₹223,000.00INR / Each
Adam Equipment CDT 32-150 Cruiser® Dual Counting Scale, 70lb / 32kg with 330lb / 150kg Remote Platform WW-55001-08 Mfr # CDT 32-150
COMPARE
32 70 Counting Scale with Remote Platform 70lb / 32kg and 330lb / 150kg
₹189,000.00INR / Each
Adam Equipment CDT 32-300 Cruiser® Dual Counting Scale, 70lb / 32kg with 660lb / 300kg Remote Platform WW-55001-09 Mfr # CDT 32-300
COMPARE
32 70 Counting Scale with Remote Platform 70lb / 32kg and 660lb / 300kg
₹189,000.00INR / Each
Adam Equipment CDT 32-600 Cruiser® Dual Counting Scale, 70lb / 32kg with 1320lb / 600kg Remote Platform WW-55001-10 Mfr # CDT 32-600
COMPARE
32 70 Counting Scale with Remote Platform 70lb / 32kg and 1320lb / 600kg
₹227,000.00INR / Each
Adam Equipment CDT 4-150 Cruiser® Dual Counting Scale, 8lb / 4kg with 330lb / 150kg Remote Platform WW-55001-11 Mfr # CDT 4-150
COMPARE
4 8 Counting Scale with Remote Platform 8lb / 4kg and 330lb / 150kg
₹189,000.00INR / Each
Adam Equipment CDT 4-32 Cruiser® Dual Counting Scale, 8lb / 4kg with 70lb / 35kg Remote Platform WW-55001-12 Mfr # CDT 4-32
COMPARE
4 8 Counting Scale with Remote Platform 8lb / 4kg and 70lb / 35kg
₹171,000.00INR / Each
Adam Equipment CDT 4-60 Cruiser® Dual Counting Scale, 8lb / 4kg with 130lb / 60kg Remote Platform WW-55001-13 Mfr # CDT 4-60
COMPARE
4 8 Counting Scale with Remote Platform 8lb / 4kg and 130lb / 60kg
₹171,000.00INR / Each
Showing 1- 14 of 14