Adam Equipment CPW Plus Industrial Scales
These versatile scales with remote mount display are ideal for a wide variety of weighing tasks
  • Battery and AC operation
  • Splashproof keypad for use in wet environments
  • Hold function lets you freeze the reading on the display
  • 3-year manufacturer's warranty
  • Color-coded keypads with backlit display provide simple and quick operation
  • Steel base offers stability for weighing tasks
  • Removable stainless steel platform for easy cleaning
  • RS-232 interface
  • Remote-mounted display
CPW Plus industrial scales from Adam Equipment provide accuracy and dependability when all you need is a weight. These durable, professional industrial scales feature a separate weight display, which is supplied with a bracket so that it can be mounted on a wall or benchtop. The weight display can be wiped down to keep clean, and the grade 304 stainless steel weighing pan can also be removed for cleaning. Use the RS-232 output to connect the weighing scale to Adam Equipment’s ATP2 Thermal Printer.
The 300x300mm base is suitable for weighing parcels, packages and other small to medium-sized items, and can be placed on a benchtop or on the floor.
All CPW Plus industrial scales come with a 3-year manufacturer’s warranty.
52,400.00 - ₹76,900.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Adam Equipment CPWplus Industrial Scale, 35kg x 0.01kg, 115 V 1190100 Mfr # CPWPLUS 35
COMPARE
35 75 115
₹56,500.00INR / Each
Adam Equipment CPWplus Industrial Scale, 75kg x 0.02kg, 115 V 1190102 Mfr # CPWPLUS 75
COMPARE
75 165 115
₹56,500.00INR / Each
Adam Equipment CPWplus Industrial Scale, 150kg x 0.05kg, 115 V 1190106 Mfr # CPWPLUS 150
COMPARE
150 330 115
₹60,600.00INR / Each
Adam Equipment CPWPLUS6-115V Industrial Scale, 6kg x 2g, 115 V 1190108 Mfr # CPWPLUS 6
COMPARE
6 13 100 to 240
₹52,400.00INR / Each
Adam Equipment CPWPLUS15-115V Industrial Scale, 15kg x 5g, 115 V 1190110 Mfr # CPWPLUS 15
COMPARE
15 33 100 to 240
₹55,400.00INR / Each
Adam Equipment CPWplus Industrial Scale, 200kg x 0.05kg, 115 V 1190112 Mfr # CPWPLUS 200
COMPARE
200 440 100 to 240
₹76,900.00INR / Each