Adam Equipment CBX Series Compact Portable Balances
Easy keypad calibration ensures accurate weighing every time
  • Backlit LCD visible in any lighting conditions
  • Simple key operation allows for easy setup and operation
  • Large stainless steel pan makes for easy cleaning
  • Level indicator and adjustable feet ensure proper balance setup
  • Auto power-off conserves battery life
The CBX portable balances offer a bright backlit display with 18 mm high digits, allowing you to operate in any degree of light. The 304-grade stainless steel top pan offers protection against most materials and liquids, and is easy to clean. With easy calibration through the keypad, the balances consistently ensure correct and accurate weighing of samples.
The balances come with a standard internal rechargeable battery which can last up to 216 hours. Low-battery indicator alerts you when you need to recharge the battery. A power adapter (included) can also be used to operate the balance. Solid ABS plastic housing is durable for most industrial and laboratory use. Additional features include parts counting with present sample sizes, tart/zero key, and overload indicator.
28,400.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Adam Equipment CBX 1202 Compact Balance, 1200g x 0.1g Readability, External Calibration w/Rechargeable Battery WW-11171-00 Mfr # CBX 1201
COMPARE
External 1200 0.1
₹28,400.00INR / Each
Adam Equipment CBX 3000 Compact Balance, 3000g x 0.5g Readability, External Calibration w/Rechargeable Battery WW-11171-01 Mfr # CBX 3001
COMPARE
External 3000 0.5
₹28,400.00INR / Each
Adam Equipment CBX 6000 Compact Balance, 6000g x 1.0g Readability, External Calibration w/Rechargeable Battery WW-11171-02 Mfr # CBX 6000
COMPARE
External 6000 1
₹28,400.00INR / Each